Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Methoxyphenoxy)-1,2-propanediol, 98+%

Catalog No. AAA168270B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA168270B 1000 g
AAA1682718 50 g
AAA1682730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA168270B Supplier Thermo Scientific Chemicals Supplier No. A168270B
Add to Cart

CAS: 93-14-1 | C10H14O4 | 198.22 g/mol

3-(2-Methoxyphenoxy)-1,2-propanediol is involved in centrally acting muscle relaxant with expectorant properties. It is often used as anesthetic during animal surgery. It is used in the treatment of coughing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Methoxyphenoxy)-1,2-propanediol is involved in centrally acting muscle relaxant with expectorant properties. It is often used as anesthetic during animal surgery. It is used in the treatment of coughing

Solubility
Soluble in water (50 mg/ml at 25°C), methanol, DMSO (40 mg/ml at 25°C), DMF, ethanol (50 mg/ml), glycerol and glycol.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-14-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00016873
InChI Key HSRJKNPTNIJEKV-UHFFFAOYNA-N
Synonym guaifenesin, guaiphenesin, glyceryl guaiacolate, glycerol guaiacolate, guaiphenesine, guaiacol glyceryl ether, methphenoxydiol, aeronesin, breonesin, bronchol
PubChem CID 3516
IUPAC Name 3-(2-methoxyphenoxy)propane-1,2-diol
SMILES COC1=CC=CC=C1OCC(O)CO

Specifications

Melting Point 80°C to 82°C
Boiling Point 215°C (19 mmHg)
Quantity 1000 g
Beilstein 2049375
Merck Index 14,4555
Solubility Information Soluble in water (50mg/ml at 25°C),methanol,DMSO (40mg/ml at 25°C),DMF,ethanol (50mg/ml),glycerol and glycol.
Formula Weight 198.22
Percent Purity ≥99%
Chemical Name or Material 3-(2-Methoxyphenoxy)-1,2-propanediol
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-222-5
RTECSNumber TY8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.