Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Methyl-4-pyrimidinyl)benzenesulfonyl chloride, Technical, Thermo Scientific™

Catalog No. CC31603CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC31603CB 250 mg Amber Glass Bottle
CC31603DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC31603CB Supplier Thermo Scientific Chemicals Supplier No. CC31603CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 465514-07-2
Molecular Formula C11H9ClN2O2S
Molecular Weight (g/mol) 268.715
InChI Key ZLLPUDLQNCNHFM-UHFFFAOYSA-N
Synonym 3-2-methylpyrimidin-4-yl benzene-1-sulfonylchloride, 3-2-methyl-4-pyrimidinyl benzenesulfonyl chloride, 3-2-methylpyrimidin-4-yl benzenesulfonyl chloride, 3-2-methylpyrimidin-4-yl benzene-1-sulfonyl chloride, 3-2-methylpyrimidin-4-yl benzenesulphonyl chloride, chloro 3-2-methylpyrimidin-4-yl phenyl sulfone, 3-2-methyl-4-pyrimidinyl phenylsulfonyl chloride, benzenesulfonylchloride, 3-2-methyl-4-pyrimidinyl, benzenesulfonyl chloride, 3-2-methyl-4-pyrimidinyl
PubChem CID 2779830
IUPAC Name 3-(2-methylpyrimidin-4-yl)benzenesulfonyl chloride
SMILES CC1=NC=CC(=N1)C2=CC(=CC=C2)S(=O)(=O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 268.72
Percent Purity 95%
Chemical Name or Material 3-(2-Methyl-4-pyrimidinyl)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.