Learn More
Bachem 3-(2-Pyridyldithio)-propionic acid-OSu
SDP
Supplier:  Bachem 4029094.0250
Encompass_Preferred
3-(2-Pyridyldithio)-propionic acid-OSu, 68181-17-9, C12H12N2O4S2, Mr 312.37, 250mg. 3-(Pyridin-2-yldisulfanyl)-propionic acid-OSu, SPDP. SPDP, important crosslinking reagent for the preparation of protein-protein and peptide-protein conjugates linked by disulfide bonds.
Catalog No. 50-260-659
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.