Learn More
Sigma Organic Chemistry 3 3-- 2-tetrahydropyran | 1G | nocas | MFCD00090526
Supplier: Sigma Organic Chemistry RNI001181G

3 3-- 2-tetrahydropyran , 1G
About this item:
MDL #: MFCD00090526
Chemical Formula: C10H16O2
Molecular Weight: 168.23
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.