Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3,3,3-Trifluoropropyl)methyldichlorosilane, 97%

Catalog No. AAL1666914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1666914 25 g
AAL1666922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1666914 Supplier Thermo Scientific Chemicals Supplier No. L1666914
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 675-62-7 | C4H7Cl2F3Si | 211.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675-62-7
Molecular Formula C4H7Cl2F3Si
Molecular Weight (g/mol) 211.08
MDL Number MFCD00039294
InChI Key OHABWQNEJUUFAV-UHFFFAOYSA-N
Synonym 3,3,3-trifluoropropyl methyldichlorosilane, 3,3,3-trifluoropropyl dichloromethylsilane, dichloro methyl 3,3,3-trifluoropropyl silane, unii-38xgw8m24k, silane, dichloromethyl 3,3,3-trifluoropropyl, dichloromethyl-3,3,3-trifluoropropylsilane, dichloromethyl 3,3,3-trifluoropropyl silane, dichloro-methyl-3,3,3-trifluoropropyl silane, 4-04-00-04169 beilstein handbook reference, ksc491g7h
PubChem CID 12666
IUPAC Name dichloro-methyl-(3,3,3-trifluoropropyl)silane
SMILES C[Si](CCC(F)(F)F)(Cl)Cl

Specifications

Boiling Point 121°C to 122°C
Quantity 25 g
Flash Point 15°C (59°F)
Refractive Index 1.385
Sensitivity Moisture sensitive
Density 1.261
UN Number UN2985
Beilstein 1744964
Formula Weight 211.09
Percent Purity 97%
Chemical Name or Material (3,3,3-Trifluoropropyl)methyldichlorosilane
Show More Show Less
Hazard Category H225-H314-H332-H335
Hazard Statement GHS H Statement
H225-H314-H318-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 211-623-4
RTECSNumber VV3531500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.