Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',4,5'-Tetramethoxypiceatannol 98.0+%, TCI America™
SDP

Catalog No. T28421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28421G 1 g
T28425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28421G Supplier TCI America Supplier No. T28421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83088-26-0
Molecular Formula C18H20O4
Molecular Weight (g/mol) 300.354
MDL Number MFCD09033675
InChI Key PTVAOGIYBMTHSN-AATRIKPKSA-N
PubChem CID 5387294
IUPAC Name 1-[(E)-2-(3,4-dimethoxyphenyl)ethenyl]-3,5-dimethoxybenzene
SMILES COC1=C(C=C(C=C1)C=CC2=CC(=CC(=C2)OC)OC)OC

Specifications

Melting Point 69°C
Color White
Quantity 1 g
Synonym 3,3′C,4,5′C-Tetramethoxy-trans-stilbene
Formula Weight 300.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,4,5′-Tetramethoxypiceatannol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.