Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetrabromo-2,2'-bithiophene 98.0+%, TCI America™
SDP

Catalog No. T269925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T269925G 25 g
T26991G 1 g
T26995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T269925G Supplier TCI America Supplier No. T269925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 125143-53-5
Molecular Formula C8H2Br4S2
Molecular Weight (g/mol) 481.84
InChI Key MOMHMPZSZNZLAK-UHFFFAOYSA-N
PubChem CID 2740586
IUPAC Name 3,5-dibromo-2-(3,5-dibromothiophen-2-yl)thiophene
SMILES C1=C(SC(=C1Br)C2=C(C=C(S2)Br)Br)Br

Specifications

Melting Point 140°C
Quantity 25 g
Formula Weight 481.84
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,5,5′-Tetrabromo-2,2′-bithiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.