Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',3″,5',5″-Tetrabromophenolphthalein 99%, Thermo Scientific™

Catalog No. AC420065000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC420065000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420065000 Supplier Thermo Scientific Chemicals Supplier No. 420065000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-62-0
Molecular Formula C20H10Br4O4
Molecular Weight (g/mol) 633.912
InChI Key OBRGVMYQZVQHGO-UHFFFAOYSA-N
Synonym tetrabromophenolphthalein, 3',3,5',5-tetrabromophenolphthalein, tetbrothalein, phenol tetrabromophthalein, unii-yz4ht20i0v, 1 3h-isobenzofuranone, 3,3-bis 3,5-dibromo-4-hydroxyphenyl, phenolphthalein, tetrabromo, 3,3-bis 3,5-dibromo-4-hydroxyphenyl isobenzofuran-1 3h-one, yz4ht20i0v, phenolphthalein, 3',3,5',5-tetrabromo
PubChem CID 65569
IUPAC Name 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC2(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 500 g
Merck Index 15, 9331
Formula Weight 633.9
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 3',3'',5',5''-Tetrabromophenolphthalein, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.