Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 3,3',5,5'-Tetramethylbenzidine Liquid Substrate

Catalog No. mp215234680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215234680 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215234680 Supplier MP Biomedicals™ Supplier No. 0215234680
Only null left
Add to Cart
Add to Cart

3,3',5,5'-Tetramethylbenzide Liquid Substrate is a chromogenic substrate for Horse Radish Peroxidase (HRP). TMB Solution is a convenient, non toxic and ready to use stable chromogen solution suitable for peroxidase and pseudo-peroxidase reactions in ELISA, it provides consistent performance and sensitivity to deliver reproducible results.

  • Ready to use 1.25 mmol/L stablized chromogen solution
  • Metabolism of the TMB substrate by HRP creates a soluble, bluish reaction product that may be quantified by measuring absorbance at 370 or 665 nm.
  • Reaction can be stopped by addition of 1 volume of 1N hydrocloric or sulfurc acid to yield a yellow reaction product that can be measured by absorbance at 450 nm (within one hour).
TRUSTED_SUSTAINABILITY

Specifications

Product Type Liquid Substrate
Format Solution
Quantity 100 mL
Storage Requirements 4°C, protect from light
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.