Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 3,3',5,5'-Tetramethylbenzidine, MP Biomedicals™

Catalog No. MP8980501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP8980501 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP8980501 Supplier MP Biomedicals™ Supplier No. 08980501
Add to Cart
Add to Cart

Reagent that is excellent for ELISA procedures/immunoassays.

Molecular Formula: C16H20N2
Molecular Weight: 240.3

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 g
Form Powder
For Use With (Application) ELISA
Content And Storage Store at room temperature. Protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.