Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5,5'-Tetramethylbenzidine, MP Biomedicals™

Catalog No. ICN15234650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 100 mg
50 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15234650 50 mL
ICN980502 10 x 100 mg
ICN15234691 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15234650 Supplier MP Biomedicals, Inc Supplier No. 0215234650
Add to Cart
Add to Cart

Excellent for ELISA procedures

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Color Colorless
Linear Formula C16H20N2
Quantity 50 mL
Formula Weight 240.3
Physical Form Liquid
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.