Learn More
3,3',5,5'-Tetramethylbenzidine, MP Biomedicals™
Catalog No. 021902771 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02190277-1 100 mg
02-190-27701 1 g
02-190-27705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02190277-1 Supplier MP Biomedicals, Inc Supplier No. 02190277.1
Add to Cart
Add to Cart
3,3',5,5'-Tetramethylbenzidine is suitable as peroxidase substrate for use in ELISA procedures.
  • The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620-650 nm.
  • The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm.
  • A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases.
  • It is useful in gel staining procedure for low levels of home-associated peroxidases and for enzyme immunoassay of horseradish peroxidase.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mg
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.