Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',5-Triiodo-L-thyronine, 95%

Catalog No. AAH3406803
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3406803 1 g
AAH34068MD 250 mg
AAH3406806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3406803 Supplier Thermo Scientific Chemicals Supplier No. H3406803
Add to Cart

CAS: 6893-02-3 | C15H12I3NO4 | 650.98 g/mol

Using video intensification fluorescence microscopy and tetramethylrhodamine (Rho)-labeled 3, 3', 5-triiodo-L-thyronine (T3),the uptake of T3 by cultured mouse fibroblasts was studied. The binding of 3, 3', 5-triiodo-L-thyronine (T3) to GH3 rat pituitary tumor cells was studied at 15 degrees C and was shown to be saturable, reversible, and stereospecific. A fluorescent derivative of the thyroid hormone 3,3'-triiodo-L-thyronine binds to cultured mouse fibroblasts; such binding is saturable. 3,3?,5-Triiodo-L-thyronine Up-regulation of Na,K-ATPase Activity and Cell Surface Expression in Alveolar Epithelial Cells Is Src Kinase- and Phosphoinositide 3-Kinase-dependent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Using video intensification fluorescence microscopy and tetramethylrhodamine (Rho)-labeled 3, 3′, 5-triiodo-L-thyronine (T3),the uptake of T3 by cultured mouse fibroblasts was studied. The binding of 3, 3′, 5-triiodo-L-thyronine (T3) to GH3 rat pituitary tumor cells was studied at 15 degrees C and was shown to be saturable, reversible, and stereospecific. A fluorescent derivative of the thyroid hormone 3,3′-triiodo-L-thyronine binds to cultured mouse fibroblasts; such binding is saturable. 3,3′,5-Triiodo-L-thyronine Up-regulation of Na,K-ATPase Activity and Cell Surface Expression in Alveolar Epithelial Cells Is Src Kinase- and Phosphoinositide 3-Kinase-dependent.

Solubility
Insoluble in water, ethanol and propylene glycol. Soluble in dilute alkalies.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Strong bases, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6893-02-3
Molecular Formula C15H12I3NO4
Molecular Weight (g/mol) 650.98
MDL Number MFCD00002593
InChI Key AUYYCJSJGJYCDS-UHFFFAOYNA-N
Synonym liothyronine, triiodothyronine, 3,3',5-triiodo-l-thyronine, liothyronin, tresitope, 3,5,3'-triiodothyronine, l-liothyronine, triothyrone, liothyroninum, liotironina
PubChem CID 5920
ChEBI CHEBI:18258
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES C1=CC(=C(C=C1OC2=C(C=C(C=C2I)CC(C(=O)O)N)I)I)O

Specifications

Melting Point 234°C to 238°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water,ethanol and propylene glycol. Soluble in dilute alkalies.
Optical Rotation +25° (c=1 in 1M HCl/Ethanol 1:2)
Formula Weight 650.97
Percent Purity 95%
Chemical Name or Material 3,3',5-Triiodo-L-thyronine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-999-3
RTECSNumber AY6750000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.