Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3,5-Trimethylcyclohexanone 98.0+%, TCI America™
SDP

Catalog No. T060225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T060225ML 25 mL
T0602500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T060225ML Supplier TCI America Supplier No. T060225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 873-94-9
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00019466
InChI Key POSWICCRDBKBMH-UHFFFAOYSA-N
Synonym 3,3,5-trimethylcyclohexanone, dihydroisophorone, cyclohexanone, 3,3,5-trimethyl, dihydro-isophorone, 3,5,5-trimethylcyclohexanone, 3,3,5-trimethyl cyclohexanone, acmc-209qk8, dsstox_cid_24996, dsstox_rid_80641, dsstox_gsid_44996
PubChem CID 13398
IUPAC Name 3,3,5-trimethylcyclohexan-1-one
SMILES CC1CC(=O)CC(C1)(C)C

Specifications

Color Colorless
Boiling Point 190°C
Quantity 25 mL
Formula Weight 140.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3,5-Trimethylcyclohexanone
EINECSNumber (3)-2391
RTECSNumber GW2185000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.