Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Bis(trimethylsilyl)biphenyl-4,4'-diyl Bis(trifluoromethanesulfonate) 98.0+%, TCI America™
SDP

Catalog No. B30475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30475G 5 g
B30471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30475G Supplier TCI America Supplier No. B30475G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 828282-80-0
Molecular Formula C20H24F6O6S2Si2
Molecular Weight (g/mol) 594.686
InChI Key FDAGZCNNWNNCEK-UHFFFAOYSA-N
PubChem CID 12182649
IUPAC Name [4-[4-(trifluoromethylsulfonyloxy)-3-trimethylsilylphenyl]-2-trimethylsilylphenyl] trifluoromethanesulfonate
SMILES C[Si](C)(C)C1=C(C=CC(=C1)C2=CC(=C(C=C2)OS(=O)(=O)C(F)(F)F)[Si](C)(C)C)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 118°C
Color White
Quantity 5 g
UN Number 3261
Synonym 3,3′-Bis(trimethylsilyl)biphenyl-4,4′-diyl Ditriflate, 4,4′-Bis(trifluoromethanesulfonyloxy)-3,3′-bis(trimethylsilyl)biphenyl
Formula Weight 594.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Bis(trimethylsilyl)biphenyl-4,4′-diyl Bis(trifluoromethanesulfonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.