Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diamino-N-methyldipropylamine 98.0+%, TCI America™
SDP

Catalog No. B082125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B082125ML 25 mL
B0821500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B082125ML Supplier TCI America Supplier No. B082125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 105-83-9
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00008217
InChI Key KMBPCQSCMCEPMU-UHFFFAOYSA-N
Synonym n,n-bis 3-aminopropyl methylamine, 3,3'-diamino-n-methyldipropylamine, bis 3-aminopropyl methylamine, n-methyliminobis propylamine, methyliminobispropylamine, methylbis 3-aminopropyl amine, 5-methyldipropylenetriamine, 1,3-propanediamine, n-3-aminopropyl-n-methyl, 3,3'-methyliminobispropylamine, n1-3-aminopropyl-n1-methylpropane-1,3-diamine
PubChem CID 7777
IUPAC Name bis(3-aminopropyl)(methyl)amine
SMILES CN(CCCN)CCCN

Specifications

Color Colorless
Boiling Point 241°C
Quantity 25 mL
UN Number 2927
Formula Weight 145.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,3′-Diamino-N-methyldipropylamine
EINECSNumber (2)-0165
RTECSNumber JL9625000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.