Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water

Catalog No. AC112090010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112090010 1 g Glass bottle
AC112090050 5 g Glass bottle
AC112090250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC112090010 Supplier Thermo Scientific Chemicals Supplier No. 112090010
Add to Cart

CAS: 868272-85-9 | C12H18Cl4N4 | 360.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3, 3'-Diaminobenzidine tetrahydrochloride hydrate
CAS 868272-85-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 300.0°C
Color Brown-Purple to Gray
Loss on Drying 10% max. (120°C)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Molecular Formula C12H18Cl4N4
Linear Formula 4HCl·xH2O
Quantity 1 g
MDL Number MFCD08273058
Beilstein 13, 340
Synonym 3,3'-diaminobenzidine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, acmc-209qbm, 3,3'-diaminobenzidine hydrate tetrahydrochloride, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se
Solubility Information Solubility in water: moderately soluble. Other solubilities: insoluble in most common organic solvents
InChI Key KJDSORYAHBAGPP-UHFFFAOYSA-N
SMILES Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1
IUPAC Name 4-(3,4-diaminophenyl)benzene-1,2-diamine;hydrate
Molecular Weight (g/mol) 360.10
PubChem CID 23191111
Formula Weight 360.11
Percent Purity 97%
Physical Form Crystalline Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.