Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Diethylthiacarbocyanine Iodide 98.0+%, TCI America™
SDP

Catalog No. D44545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44545G 5 g
D44541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44545G Supplier TCI America Supplier No. D44545G

Chemical Identifiers

CAS 905-97-5
Molecular Formula C21H22IN2S2+
Molecular Weight (g/mol) 493.445
MDL Number MFCD00011956
InChI Key VZBILKJHDPEENF-UHFFFAOYSA-N
Synonym 3,3'-diethylthiacarbocyanine iodide
PubChem CID 129893628
IUPAC Name (2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzothiazole;hydroiodide
SMILES CCN1C2=CC=CC=C2SC1=CC=CC3=[N+](C4=CC=CC=C4S3)CC.I

Specifications

Melting Point 268°C
Quantity 5 g
Formula Weight 492.44
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diethylthiacarbocyanine Iodide
RTECSNumber DL7061500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.