Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dihydroxybenzidine 99.0+%, TCI America™
SDP

Catalog No. D231225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D231225G 25 g
D23125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D231225G Supplier TCI America Supplier No. D231225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2373-98-0
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD00039149
InChI Key ZGDMDBHLKNQPSD-UHFFFAOYSA-N
Synonym Benzidine-3,3′C-diol, 4,4′C-Diamino-3,3′C-dihydroxybiphenyl
PubChem CID 16918
IUPAC Name 4,4'-diamino-[1,1'-biphenyl]-3,3'-diol
SMILES NC1=CC=C(C=C1O)C1=CC=C(N)C(O)=C1

Specifications

Melting Point 292°C
Quantity 25 g
Formula Weight 216.24
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dihydroxybenzidine
RTECSNumber DV4900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.