Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Dimethyl-2-oxobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. D360925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D360925G 25 g
D36095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D360925G Supplier TCI America Supplier No. D360925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 815-17-8
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00154352
InChI Key IAWVHZJZHDSEOC-UHFFFAOYSA-N
Synonym 3,3-dimethyl-2-oxobutyric acid, trimethylpyruvic acid, butanoic acid, 3,3-dimethyl-2-oxo, pyruvic acid, trimethyl, glyoxylic acid, tert-butyl, unii-29cl615clh, 3,3-dimethyl-2-oxo-butanoic acid, butyric acid, 3,3-dimethyl-2-oxo, tert-butylglyoxylic acid, trimethlpyruvic acid
PubChem CID 13150
IUPAC Name 3,3-dimethyl-2-oxobutanoic acid
SMILES CC(C)(C)C(=O)C(=O)O

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 g
UN Number 3265
Formula Weight 130.14
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-2-oxobutyric Acid
EINECSNumber (2)-1503
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.