Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dimethylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. D143025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D143025ML 25 mL
D14305ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D143025ML Supplier TCI America Supplier No. D143025ML
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 612-75-9
Molecular Formula C14H14
Molecular Weight (g/mol) 182.27
MDL Number MFCD00008534
InChI Key GVEDOIATHPCYGS-UHFFFAOYSA-N
Synonym 3,3'-dimethylbiphenyl, 3,3'-dimethyl-1,1'-biphenyl, 3,3'-ditolyl, m,m'-bitolyl, 1,1'-biphenyl, 3,3'-dimethyl, 1-methyl-3-3'-methylphenyl benzene, 1,1'-biphenyl,3,3'-dimethyl, 1-methyl-3-3-methylphenyl benzene, pubchem3146, 3,3-dimethyldiphenyl
PubChem CID 11931
IUPAC Name 3,3'-dimethyl-1,1'-biphenyl
SMILES CC1=CC(=CC=C1)C1=CC(C)=CC=C1

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 mL
Formula Weight 182.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3′-Dimethylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.