Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Diphenyl-1-propanol 98.0+%, TCI America™
SDP

Catalog No. D276025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D276025G 25 g
D27605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D276025G Supplier TCI America Supplier No. D276025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20017-67-8
Molecular Formula C15H16O
Molecular Weight (g/mol) 212.29
MDL Number MFCD00002930
InChI Key IDCXQMVSIIJUEH-UHFFFAOYSA-N
PubChem CID 29908
IUPAC Name 3,3-diphenylpropan-1-ol
SMILES OCCC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 208°C
Quantity 25 g
Formula Weight 212.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Diphenyl-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.