Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Diphenylpropionitrile 97.0+%, TCI America™
SDP

Catalog No. D259025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D259025G 25 g
D25905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D259025G Supplier TCI America Supplier No. D259025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2286-54-6
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00129747
InChI Key INERKLNEVAZSCI-UHFFFAOYSA-N
Synonym 2-Cyano-1,1-diphenylethane
PubChem CID 75295
IUPAC Name 3,3-diphenylpropanenitrile
SMILES C1=CC=C(C=C1)C(CC#N)C2=CC=CC=C2

Specifications

Melting Point 86°C
Boiling Point 174°C
Quantity 25 g
Formula Weight 207.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Diphenylpropionitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.