Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Diphenylpropyl Bromide 98.0+%, TCI America™
SDP

Catalog No. D278425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D278425G 25 g
D27845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D278425G Supplier TCI America Supplier No. D278425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20017-68-9
Molecular Formula C15H15Br
Molecular Weight (g/mol) 275.19
MDL Number MFCD00044829
InChI Key SLHSRCBFPHCSGL-UHFFFAOYSA-N
Synonym 1-bromo-3,3-diphenylpropane, 3,3-diphenylpropyl bromide, 3-bromo-1,1-diphenylpropane, 3-bromopropane-1,1-diyl dibenzene, 3-bromo-1-phenylpropyl benzene, 3,3-diphenylpropylbromide, 1,1'-3-bromopropylidene bisbenzene, benzene, 1,1'-3-bromopropylidene bis, acmc-1cjoi, 3,3-diphenyl-1-propylbromide
PubChem CID 88333
IUPAC Name (3-bromo-1-phenylpropyl)benzene
SMILES BrCCC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 40°C
Color White
Boiling Point 145°C
Quantity 25 g
Formula Weight 275.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Diphenylpropyl Bromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.