Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 3,3'-Dipropylthiadicarbocyanine Iodide, Fluorogenic Probe, >98% Purity, Powder, 100 mg

Catalog No. 502705224
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5224 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5224 Supplier Abcam Supplier No. AB145407100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogenic probe used to measure changes in transmembrane potential or configuration induced by membrane-altering agents.

  • Purity: >98%
  • Molecular Weight: 546.53 g/mol
  • Applications: Fluorogenic probe used to detect and measure changes in transmembrane potential or configuration induced by membrane-altering agents
  • Form: Powder
  • Storage: Store at ambient temperature

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Transmembrane Potential
Form Powder
Shipping Condition Ambient
Detection Method Fluorescence
Content And Storage Store at ambient temperature.
Substrate Properties Chemical Substrate
Formulation 100% 3,3'-Dipropylthiadicarbocyanine iodide
Quantity 100 mg
For Use With (Application) Detecting and measuring changes in transmembrane potential or configuration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.