Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 3,3'-Dipropylthiadicarbocyanine Iodide, Fluorogenic Probe, Powder, 1 g

Catalog No. 502705223
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5223 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5223 Supplier Abcam Supplier No. AB1454071G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogenic probe for detecting and measuring changes in transmembrane potential or configuration.

  • Purity: >98%
  • Molecular Weight: 546.53 g/mol
  • Chemical Form: Powder
  • Application: Fluorogenic probe used to detect and measure changes in transmembrane potential or configuration induced by membrane altering agents
  • Composition: 100% 3,3'-Dipropylthiadicarbocyanine iodide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Transmembrane Potential
Label or Dye Fluorogenic Probe
Shipping Condition Ambient
Form Powder
Detection Method Fluorescence
Buffer 100% 3,3'-Dipropylthiadicarbocyanine iodide
Content And Storage Store at ambient temperature.
Formulation 100% 3,3'-Dipropylthiadicarbocyanine iodide
Quantity 1 g
For Use With (Application) Detecting and measuring changes in transmembrane potential, detecting and measuring membrane configuration changes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.