Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Iminodipropionic Acid 98.0+%, TCI America™
SDP

Catalog No. I08235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08235G 5 g
I08231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08235G Supplier TCI America Supplier No. I08235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 505-47-5 | C6H11NO4 | 161.16 g/mol

Chemical Identifiers

CAS 505-47-5
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD01311684
InChI Key TXPKUUXHNFRBPS-UHFFFAOYSA-N
PubChem CID 10456
IUPAC Name 3-[(2-carboxyethyl)amino]propanoic acid
SMILES OC(=O)CCNCCC(O)=O

Specifications

Melting Point 156°C
Color White
Quantity 5 g
Formula Weight 161.16
Percent Purity ≥98.0% (GC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Iminodipropionic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.