Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Iminodipropionitrile 98.0+%, TCI America™
SDP

Catalog No. I0010100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0010100ML 100 mL
I001025ML 25 mL
I0010500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0010100ML Supplier TCI America Supplier No. I0010100ML
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 111-94-4
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.159
MDL Number MFCD00001956
InChI Key SBAJRGRUGUQKAF-UHFFFAOYSA-N
Synonym 3,3'-iminodipropionitrile, bis 2-cyanoethyl amine, propanenitrile, 3,3'-iminobis, iminodipropanenitrile, idpn, 3,3'-iminobispropanenitrile, bbce, 3,3'-iminodipropanenitrile, n,n-bis 2-cyanoethyl amine, bis beta-cyanoethyl amine
PubChem CID 8149
IUPAC Name 3-(2-cyanoethylamino)propanenitrile
SMILES C(CNCCC#N)C#N

Specifications

Color Yellow
Boiling Point 205°C
Quantity 100 mL
Formula Weight 123.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3′-Iminodipropionitrile
EINECSNumber (2)-1530
RTECSNumber UG2975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.