Learn More
Sigma Organic Chemistry 3-3-methoxyphenyl thiom | 100mg | MFCD06797782
Supplier: Sigma Organic Chemistry CDS009236100MG

3-3-methoxyphenyl thiom , 100mg
About this Item:
MDL #: MFCD06797782
Molecular Weight: 209.31
UNSPSC Code: 12352200
Chemical Formula: C11H15NOS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.