Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3-Pyridyl)-D-alanine, 95%

Catalog No. AAH63132MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63132MD 250 mg
AAH6313203 1 g
AAH6313206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63132MD Supplier Thermo Scientific Chemicals Supplier No. H63132MD
Only null left
Add to Cart

CAS: 70702-47-5 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70702-47-5
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00079673
InChI Key DFZVZEMNPGABKO-UHFFFAOYNA-N
Synonym 3-3-pyridyl-d-alanine, d-3-pyridylalanine, d-3-3-pyridyl-alanine, h-3-d-pal-oh, 3'-pyridyl-d-ala, r-2-amino-3-pyridin-3-yl propanoic acid, 2r-2-amino-3-pyridin-3-yl propanoic acid, 2r-2-amino-3-3-pyridyl propanoic acid, 3-aza-d-phenylalanine, 3-aza-l-phenylalanine
PubChem CID 1268144
SMILES NC(CC1=CN=CC=C1)C(O)=O

Specifications

Color White
Beilstein 4351584
IUPAC Name 2-amino-3-(pyridin-3-yl)propanoic acid
Quantity 250 mg
Formula Weight 166.18
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-(3-Pyridyl)-D-alanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.