Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[3-(Trifluoromethyl)phenyl]-1-propanol 98.0+%, TCI America™
SDP

Catalog No. T30951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30951G 1 g
T30955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30951G Supplier TCI America Supplier No. T30951G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78573-45-2
Molecular Formula C10H11F3O
Molecular Weight (g/mol) 204.19
MDL Number MFCD08706408
InChI Key QWXKQVIMGVVIBX-UHFFFAOYSA-N
Synonym 3-3-trifluoromethyl phenyl propan-1-ol, 3-3'-trifluoromethyl phenyl propanol, 3-3-trifluoromethyl phenyl-1-propanol, 3-trifluoromethyl benzenepropanol, 3-3-trifluoromethyl-phenyl-propan-1-ol, 3-3'-trifluoromethylphenyl propanol, benzenepropanol, 3-trifluoromethyl, pubchem13744, benzenepropanol,3-trifluoromethyl
PubChem CID 18755618
ChEBI CHEBI:48528
IUPAC Name 3-[3-(trifluoromethyl)phenyl]propan-1-ol
SMILES OCCCC1=CC(=CC=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
Formula Weight 204.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-[3-(Trifluoromethyl)phenyl]-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.