Learn More
Accuflow Systems Saint-Gobain Pure-Fit TCL Tube Clamp, PVDF, 3/4 to 2 In
Supplier:  Accuflow Systems PFTCL150
Encompass_Preferred
The Pure-Fit TCL large tube clamp is a PVDF pinch clamp engineered to provide positive fluid stoppage for large-diameter, thick-wall, reinforced, and multilayer process tubing in bioprocessing and pharmaceutical fluid systems. Its smooth, low-profile outer body reduces dead volume and tubing stress, and it can be operated in the flow position to pinch tubing for complete shutoff.
  • Made from PVDF (polyvinylidene fluoride) for chemical resistance.
  • Compatible with tubing outer diameters 3/4 to 2 in (19.05 to 50.8 mm).
  • Provides positive fluid stoppage when engaged, usable in flow position.
  • Smooth, low-profile body to minimize dead legs and tubing stress.
  • Suitable for thick-wall, reinforced, and multilayer tubing.
  • Typically supplied in packs of 25, pack quantity may vary by seller.
Catalog No. NC2312811