Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. T234525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T234525G 25 g
T23451G 1 g
T23455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T234525G Supplier TCI America Supplier No. T234525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 329214-79-1
Molecular Formula C11H16BNO2
Molecular Weight (g/mol) 205.06
MDL Number MFCD03084758
InChI Key XEMDFESAXKSEGI-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-pyridineboronic acid pinacol ester, pyridine-3-boronic acid pinacol ester, 3-pyridylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-3-boronic acid, pinacol ester, 2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
PubChem CID 2734656
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC=CN=C1

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 25 g
Formula Weight 205.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.