Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5,6-Tetrachlorofluorescein, TCI America™
SDP

Catalog No. T00661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T00661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T00661G Supplier TCI America Supplier No. T00661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6262-21-1
Molecular Formula C20H8Cl4O5
Molecular Weight (g/mol) 470.08
MDL Number MFCD00010030
InChI Key QCPFFGGFHNZBEP-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrachlorofluorescein, tetrachlorofluorescein, unii-o73wzt10by, o73wzt10by, 4,5,6,7-tetrachloro-3',6'-dihydroxy-3h-spiro 2-benzofuran-1,9'-xanthen-3-one, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 4,5,6,7-tetrachloro-3',6'-dihydroxy, 4,5,6,7-tetrachloro-3',6'-dihydroxyspiro isobenzofuran-1 3h ,9'-9h xanthene-3-one, fluorescein, tetrachloro, 4,6,7-tetrachloro-fluorescein
PubChem CID 80424
IUPAC Name 4,5,6,7-tetrachloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1

Specifications

Quantity 1 g
Formula Weight 470.08
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5,6-Tetrachlorofluorescein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.