accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6262-21-1

IUPAC Name: 4,5,6,7-tetrachloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 4,5,6,7-tetrachloro-3',6'-dihydroxyspiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 470.08
Molecular Formula: C20H8Cl4O5
InChi Key: QCPFFGGFHNZBEP-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)