Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4,5-Dimethyl-2-thazolyl)-2,5-diphenyl-2H-tetrazolium bromide, 99.10%, MP Biomedicals™

Catalog No. ICN19459291 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19459291 1 g
ICN19459280 100 mg
ICN19459290 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19459291 Supplier MP Biomedicals, Inc Supplier No. 0219459291
Add to Cart
Add to Cart

Chemical Identifiers

CAS 298-93-1
Molecular Formula C18H16BrN5S
Molecular Weight (g/mol) 414.33
MDL Number MFCD00011964,MFCD00066662
InChI Key AZKSAVLVSZKNRD-UHFFFAOYSA-M
Synonym thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i
PubChem CID 64965
ChEBI CHEBI:53233
IUPAC Name 2-(4,5-dimethyl-1,3-thiazol-2-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium bromide
SMILES [Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Orange-Yellow
Quantity 1 g
Formula Weight 414.32
Percent Purity 0.991
Physical Form Crystalline Powder
Chemical Name or Material 3-(4, 5-Dimethyl-2-thazolyl)-2, 5-diphenyl-2H-tetrazolium bromide
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.