Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trihydroxybenzoic acid, 98%

Catalog No. AAB248870B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB248870B 1000 g
AAB2488730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB248870B Supplier Thermo Scientific Chemicals Supplier No. B248870B
Only null left
Add to Cart

CAS: 149-91-7 | C7H6O5 | 170.12 g/mol

Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Organic acid is used as a standard for determining phenols in the Folin-Ciocalteau assay3,4,5-Trihydroxybenzoic acid is used as a cyclooxygease inhibitor substance, which is found in plants. In analytical chemistry, it is used as a standard to determine phenol content in various analytes. It acts as a starting material for the preparation of psychedelic alkaloid mescaline. Further, it is used to produce polyesters based on phloretic acid and gallic acid. In addition to this, it is used in the preparation of its esters such as methyl gallate, ethyl gallate, propyl gallate, octyl gallate and doceyl gallate.

Solubility
Soluble in water, alcohol, ether, glycerol and acetone. Insoluble in benzene, chloroform and petroleum ether.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents, strong bases, acid anhydrides, acid chlorides, metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-91-7
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002510
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-N
Synonym gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech
PubChem CID 370
ChEBI CHEBI:30778
IUPAC Name 3,4,5-trihydroxybenzoic acid
SMILES OC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Melting Point ∼251°C (decomposition)
Density 1.694
Quantity 1000 g
Beilstein 2050274
Sensitivity Hygroscopic; Light sensitive
Merck Index 144345
Solubility Information Soluble in water,alcohol,ether,glycerol and acetone. Insoluble in benzene,chloroform and petroleum ether.
Formula Weight 170.12
Percent Purity 0.99
Chemical Name or Material 3,4,5-Trihydroxybenzoic acid, anhydrous
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-749-9
RTECSNumber LW7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.