Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trihydroxybenzoic acid monohydrate, 99%

Catalog No. AAA163030B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA163030B 1000 g
AAA1630330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA163030B Supplier Thermo Scientific Chemicals Supplier No. A163030B
Add to Cart

CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol

This chemical is used for the determination of cerium, titanium, phosphate and nitrite with the spectrophotometry. It's also used in the manufacture of thiazine dyes, oxazine dyes, drugs, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This chemical is used for the determination of cerium, titanium, phosphate and nitrite with the spectrophotometry. It′s also used in the manufacture of thiazine dyes, oxazine dyes, drugs, etc.

Solubility
It is soluble in alcohol, ether, glycerol, acetone negligible in benzene, chloroform, petroleum ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Light sensitive. Keep away from Strong oxidizing agents, Strong bases, Acid chlorides, Acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5995-86-8
Molecular Formula C7H5O5
Molecular Weight (g/mol) 169.11
MDL Number MFCD00149098
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-M
Synonym gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0
PubChem CID 24721416
IUPAC Name 3,4,5-trihydroxybenzoic acid;hydrate
SMILES OC1=CC(=CC(O)=C1O)C([O-])=O

Specifications

Melting Point 250°C (decomposition)
Quantity 1000 g
Sensitivity Light sensitive
Merck Index 144345
Solubility Information It is soluble in alcohol,ether,glycerol,acetone negligible in benzene,chloroform,petroleum ether.
Formula Weight 188.14 (170.12 Anhydrous)
Percent Purity 0.99
Chemical Name or Material 3,4,5-Trihydroxybenzoic acid monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-749-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.