Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxybenzamide 98.0+%, TCI America™
SDP

Catalog No. T071810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T071810G 10 g
T071825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T071810G Supplier TCI America Supplier No. T071810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3086-62-2
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.217
MDL Number MFCD00017129
InChI Key GGNMTJKRHHLJHH-UHFFFAOYSA-N
Synonym benzamide, 3,4,5-trimethoxy, 3,5-trimethoxybenzamide, cbmicro_005198, acmc-209hj6, 4-10-00-02020 beilstein handbook reference, wln: zvr co1 do1 eo1, 3,4,5-trimethoxybenzamide, 3,4,5-trimethoxy benzamide
PubChem CID 18334
IUPAC Name 3,4,5-trimethoxybenzamide
SMILES COC1=CC(=CC(=C1OC)OC)C(=O)N

Specifications

Melting Point 180°C
Color White
Quantity 10 g
Formula Weight 211.22
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxybenzamide
RTECSNumber CV6125000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.