Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxyphenylacetonitrile 97.0+%, TCI America™
SDP

Catalog No. T192725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T192725G 25 g
T19275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T192725G Supplier TCI America Supplier No. T192725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13338-63-1
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00001912
InChI Key ACFJNTXCEQCDBX-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyphenylacetonitrile, 2-3,4,5-trimethoxyphenyl acetonitrile, 3,4,5-trimethoxybenzyl cyanide, 3,4,5-trimethoxybenzylnitrile, 3,4,5-trimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenylacetylnitryl, 3,4,5-trimethoxyphenyl acetonitrile, acetonitrile, 3,4,5-trimethoxyphenyl, 2-3,4,5-trimethoxyphenyl ethanenitrile
PubChem CID 25887
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetonitrile
SMILES COC1=CC(CC#N)=CC(OC)=C1OC

Specifications

Color White
Quantity 25 g
UN Number 3439
Formula Weight 207.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxyphenylacetonitrile
RTECSNumber AM2475000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.