Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,6-Tri-O-benzyl-2-deoxy-D-galactopyranose 98.0+%, TCI America™
SDP

Catalog No. T1932100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1932100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1932100MG Supplier TCI America Supplier No. T1932100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protected sugar building block with >98.0% HPLC purity.

Chemical Identifiers

CAS 94189-64-7
Molecular Formula C27H30O5
Molecular Weight (g/mol) 434.53
MDL Number MFCD06797169
InChI Key PDGHLARNGSMEJE-UHFFFAOYNA-N
Synonym 2-Deoxy-D-galactopyranose 3,4,6-Tribenzyl Ether
PubChem CID 11122952
IUPAC Name 4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
SMILES OC1CC(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(COCC2=CC=CC=C2)O1

Specifications

Color White
Quantity 100 mg
Formula Weight 434.53
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 3,4,6-Tri-O-benzyl-2-deoxy-D-galactopyranose
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.