Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene 95.0+%, TCI America™
SDP

Catalog No. B1366100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1366100MG 100 mg
B13661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1366100MG Supplier TCI America Supplier No. B1366100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84-19-5
Molecular Formula C22H22O4
Molecular Weight (g/mol) 350.414
MDL Number MFCD00063286
InChI Key YWLLGDVBTLPARJ-LWKKFVLGSA-N
Synonym Dienestrol Diacetate
PubChem CID 24884313
IUPAC Name [4-[(2Z,4E)-4-(4-acetyloxyphenyl)hexa-2,4-dien-3-yl]phenyl] acetate
SMILES CC=C(C1=CC=C(C=C1)OC(=O)C)C(=CC)C2=CC=C(C=C2)OC(=O)C

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 100 mg
Formula Weight 350.41
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Bis(4-acetoxyphenyl)-2,4-hexadiene
RTECSNumber SL0590000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.