Learn More
3,4-Dibromotoluene, 98+%
Description
It can be used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It can be used in organic synthesis.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | -10°C |
| Density | 1.85 |
| Boiling Point | 251°C |
| Flash Point | 116°C (241°F) |
| Refractive Index | 1.6 |
| Quantity | 25 g |
| Beilstein | 1931706 |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 249.94 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.