Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dibromotoluene 98.0+%, TCI America™
SDP

Catalog No. D246225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D246225G 25 g
D24625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D246225G Supplier TCI America Supplier No. D246225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60956-23-2
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00079744
InChI Key LDCPXNOCWDGYIU-UHFFFAOYSA-N
Synonym 3,4-dibromotoluene, benzene, 1,2-dibromo-4-methyl, 3,4 dibromo toluene, benzene,1,2-dibromo-4-methyl, 3,4-dibromtoluol, 3,4 dibromotoluene, pubchem3930, acmc-1b64t, ksc493e6j
PubChem CID 34679
IUPAC Name 1,2-dibromo-4-methylbenzene
SMILES CC1=CC(=C(C=C1)Br)Br

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 241°C
Quantity 25 g
Formula Weight 249.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dibromotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.