Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichloro-1-butene, 98%, Thermo Scientific™

Catalog No. AC170301000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC170301000 100 mL
AC170305000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170301000 Supplier Thermo Scientific Chemicals Supplier No. 170301000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 760-23-6
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.992
MDL Number MFCD00000947
InChI Key XVEASTGLHPVZNA-UHFFFAOYSA-N
Synonym 3,4-dichloro-1-butene, 1-butene, 3,4-dichloro, 1,2-dichloro-3-butene, 3,4-dichlorobutene-1, 1,2-dichlorobut-3-ene, 3,4-dichlorobutene-1 racemic mixture, 1,2-dichloro-3-butene racemic mixture, 1-butene, 3,4-dichloro-, +-, 1-butene, 3,4-dichloro-racemic mixture, dsstox_cid_2113
PubChem CID 12971
IUPAC Name 3,4-dichlorobut-1-ene
SMILES C=CC(CCl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.15g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula ClCH2CH(Cl)CH=CH2
Refractive Index 1.4648 to 1.4668 (20°C, 589nm)
Quantity 100 mL
Formula Weight 125
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichloro-1-butene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Toxic if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-079-0
RTECSNumber EM4740000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.