Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichlorobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. D0332250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0332250G 250 g
D033225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0332250G Supplier TCI America Supplier No. D0332250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6287-38-3
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003351
InChI Key ZWUSBSHBFFPRNE-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dichloro, unii-i6z632wsj5, ccris 6015, pubchem8230, 3,4-dichlorobenzaldehye, 3,4-dichlorobenzaldhyde, 3,4 dichlorobenzaldehyde, 3,4-dichlorobenzaidehyde, 4,3-dichlorobenzaldehyde, dsstox_cid_4971
PubChem CID 22710
IUPAC Name 3,4-dichlorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)Cl)Cl

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 248°C
Quantity 250 g
UN Number 1759
Formula Weight 175.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.