Learn More
Apexbio Technology LLC Zymosan A 58856-93-2 50mg
SDP
Supplier:  Apexbio Technology LLC A993450
Encompass_Preferred
Zymosan A (CAS 58856-93-2) is a polysaccharide compound derived from the cell wall of Saccharomyces cerevisiae It exerts its biological effects by interacting with pattern recognition receptors notably Toll-like receptor 2 (TLR2) and Dectin-1 on immune cells Engagement of these receptors by Zymosan A leads to the activation of intracellular signaling pathways resulting in the production of cytokines and inflammatory mediators This immunostimulatory property makes Zymosan A a valuable tool in studies of innate immune responses inflammation and the evaluation of anti-inflammatory drug candidates
Catalog No. 50-000-95908
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.