Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4'-Dichloropropiophenone 98.0+%, TCI America™
SDP

Catalog No. D336525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D336525G 25 g
D33655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D336525G Supplier TCI America Supplier No. D336525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6582-42-9
Molecular Formula C9H8Cl2O
Molecular Weight (g/mol) 203.06
MDL Number MFCD00000554
InChI Key AJUAASWQUWIMHM-UHFFFAOYSA-N
Synonym 3',4'-dichloropropiophenone, 1-3,4-dichlorophenyl propan-1-one, 1-3,4-dichlorophenyl-1-propanone, 1-propanone, 1-3,4-dichlorophenyl, 1-propanone,1-3,4-dichlorophenyl, enamine_003892, acmc-1b5az, 3,4-dichloro propiophenone, 1-3,4-dichlorophenyl-1-propanone #
PubChem CID 599598
IUPAC Name 1-(3,4-dichlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 247°C
Quantity 25 g
Formula Weight 203.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dichloropropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.