Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3',4'-Dichlorosalicylanilide 98.0+%, TCI America™
SDP

Catalog No. D10771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10771G 1 g
D10775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10771G Supplier TCI America Supplier No. D10771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24448-73-5
Molecular Formula C13H9Cl2NO2
Molecular Weight (g/mol) 282.12
MDL Number MFCD00020023
InChI Key XWZCOIHEGFREJR-UHFFFAOYSA-N
PubChem CID 352402
IUPAC Name N-(3,4-dichlorophenyl)-2-hydroxybenzamide
SMILES C1=CC=C(C(=C1)C(=O)NC2=CC(=C(C=C2)Cl)Cl)O

Specifications

Melting Point 214°C
Quantity 1 g
Formula Weight 282.12
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dichlorosalicylanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.