Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Difluorobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. D234125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D234125G 25 g
D23415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D234125G Supplier TCI America Supplier No. D234125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64248-62-0
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.11
MDL Number MFCD00011666
InChI Key BTBFCBQZFMQBNT-UHFFFAOYSA-N
Synonym benzonitrile, 3,4-difluoro, 4-cyano-1,2-difluorobenzene, 3,4-difluoro benzonitrile, 3,4-difluorobenzenecarbonitrile, pubchem1562, acmc-209nkz, 3,4 difluorobenzonitrile, 3,4-difluoro-benzonitrile, ksc355i7l, 4-fluoro-3-fluorobenzonitrile
PubChem CID 587203
IUPAC Name 3,4-difluorobenzonitrile
SMILES FC1=CC=C(C=C1F)C#N

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 95°C
Quantity 25 g
UN Number 3439
Formula Weight 139.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Difluorobenzonitrile
EINECSNumber (3)-4305
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.